Cadmium ≥99.95%, shot

Supplier: STREM CHEMICALS INC MS
Danger

93-4850-250G 93-4850-50G
100199-118EA 56.62 USD
100199-118 100209-132
Cadmium ≥99.95%, shot
Cadmium
Formula: Cd
MW: 112.41 g/mol
Boiling Pt: 765 °C
Melting Pt: 320.9 °C
Density: 8.642
MDL Number: MFCD00010914
CAS Number: 7440-43-9
EINECS: 231-152-8
UN: 2570
ADR: 6.1,I
Merck Index: 12,01649

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR