Cadmium ≥99.999%, powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4848-10G
93-4848-50G
100207-478EA
1607.88
USD
100207-478
100199-572
Cadmium ≥99.999%, powder
Cadmium
Formula:
Cd MW: 112.41 g/mol Boiling Pt: 765 °C Melting Pt: 320.9 °C Density: 8.642 |
MDL Number:
MFCD00010914 CAS Number: 7440-43-9 EINECS: 231-152-8 UN: 2570 ADR: 6.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...