Cadmium(II) fluoride ≥99.98% (Cd basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Cadmium fluoride
93-4810-10G
93-4810-50G
100204-342EA
110.4
USD
100204-342
100200-346
Cadmium(II) fluoride ≥99.98% (Cd basis)
Cadmium (II) fluoride
Formula:
CdF₂ MW: 150.41 g/mol Boiling Pt: 1758 °C Melting Pt: 1100 °C Density: 6.64 |
MDL Number:
MFCD00010919 CAS Number: 7790-79-6 EINECS: 232-222-0 UN: 2570 ADR: 6.1,III Merck Index: 12,01655 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...