Silver ≥99.999%, shot
Supplier: STREM CHEMICALS INC MS
93-4763-50G
93-4763-10G
100206-036EA
481.23
USD
100206-036
100202-148
Silver ≥99.999%, shot
Silver
Formula:
Ag MW: 107.87 g/mol Boiling Pt: 2212 °C Melting Pt: 960.5 °C Density: 10.49 Storage Temperature: Ambient |
MDL Number:
MFCD00003397 CAS Number: 7440-22-4 EINECS: 231-131-3 Merck Index: 12,08647 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...