Silver carbonate ≥99% (Ag basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Silver(I) carbonate
93-4706-50G
93-4706-10G
100199-106EA
110.4
USD
100199-106
100197-864
Silver carbonate ≥99% (Ag basis)
Silver carbonate
Formula:
CAg₂O₃ MW: 275.75 g/mol Melting Pt: 218 °C Density: 6,077 Storage Temperature: Ambient |
MDL Number:
MFCD00003403 CAS Number: 534-16-7 EINECS: 208-590-3 Merck Index: 12,08650 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...