Niobium(V) fluoride ≥99.5%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4121-25G
93-4121-5G
100198-380EA
110.4
USD
100198-380
100208-172
Niobium(V) fluoride ≥99.5%
Niobium (V) fluoride
Formula:
F₅Nb MW: 187.9 g/mol Boiling Pt: ~220 °C Melting Pt: 72 °C Density: 3.92 |
MDL Number:
MFCD00011129 CAS Number: 7783-68-8 EINECS: 232-020-2 UN: 1759 ADR: 8,I Merck Index: 12,06650 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...