Zirconium tetrafluoride ≥99.9% (Zr basis)
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Zirconium (IV) fluoride
93-4024-10G
93-4024-50G
100200-564EA
96.25
USD
100200-564
100197-232
Zirconium tetrafluoride ≥99.9% (Zr basis)
Zirconium tetrafluoride
Formula:
F₄Zr MW: 167.22 g/mol Melting Pt: ~600 °C (subl.) Density: 4.43 |
MDL Number:
MFCD00011307 CAS Number: 7783-64-4 EINECS: 232-018-1 UN: 3260 ADR: 8,II Merck Index: 12,10308 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...