Zirconium tetrabromide ≥98% (99+%-Zr)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-4021-5G
93-4021-25G
100200-144EA
393.48
USD
100200-144
100202-120
Zirconium tetrabromide ≥98% (99+%-Zr)
Zirconium tetrabromide
Formula:
Br₄Zr MW: 410.84 g/mol Melting Pt: 450 °C (15 atm) Density: 4.201 |
MDL Number:
MFCD00049637 CAS Number: 13777-25-8 EINECS: 237-417-4 UN: 1759 ADR: 8,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...