Yttrium ≥99.9%, chips

Supplier: STREM CHEMICALS INC MS
Warning

93-3941-5G 93-3941-25G
100200-562EA 96.25 USD
100200-562 100209-114
Yttrium ≥99.9%, chips
Yttrium
Formula: Y
MW: 88.91 g/mol
Boiling Pt: 3380 °C
Melting Pt: 1522 °C
Density: 4,47
MDL Number: MFCD00011468
CAS Number: 7440-65-5
EINECS: 231-174-8
Merck Index: 12,10240

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR