Strontium oxalate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-3815-100G
93-3815-500G
100196-638EA
62.28
USD
100196-638
100210-380
Strontium oxalate ≥95%
Strontium oxalate
Formula:
C₂O₄Sr MW: 175.64 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00050803 CAS Number: 814-95-9 EINECS: 212-415-6 UN: 3288 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...