Arsenic(III) bromide ≥99.9% (As basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-3308-50G
100210-758EA 399.14 USD
100210-758
Arsenic(III) bromide ≥99.9% (As basis)
Arsenic (III) bromide
Formula: AsBr₃
MW: 314.63 g/mol
Boiling Pt: 221 °C
Melting Pt: 32.8 °C
Density: 3.54
MDL Number: MFCD00014164
CAS Number: 7784-33-0
EINECS: 232-057-4
UN: 1555
ADR: 6.1,II
Merck Index: 12,00840

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR