Germanium(IV) bromide ≥99.99% (Ge basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-3225-10G
100202-092EA 510.95 USD
100202-092
Germanium(IV) bromide ≥99.99% (Ge basis), PURATREM
Germanium (IV) bromide
Formula: Br₄Ge
MW: 392.23 g/mol
Boiling Pt: 186.5 °C
Melting Pt: 26.1 °C
Density: 3.132
MDL Number: MFCD00016115
CAS Number: 13450-92-5
EINECS: 236-612-1
UN: 1760
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR