Germanium(IV) oxide ≥99.999% (Ge basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Germanium dioxide

93-3221-5G 93-3221-25G
100201-104EA 271.75 USD
100201-104 100207-832
Germanium(IV) oxide ≥99.999% (Ge basis), PURATREM
Germanium (IV) oxide
Formula: GeO₂
MW: 104.61 g/mol
Melting Pt: 1111…1119 °C
Density: 4.228
Storage Temperature: Ambient
MDL Number: MFCD00011030
CAS Number: 1310-53-8
EINECS: 215-180-8
Merck Index: 12,04417

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR