Gallium(III) chloride, anhydrous ≥99.999%, fused lump, PURATREM
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Gallium trichloride
93-3141-100G
93-3141-25G
93-3141-5G
100208-714EA
222.22
USD
100208-714
100199-558
100199-070
Gallium(III) chloride, anhydrous ≥99.999%, fused lump, PURATREM
Gallium (III) chloride
Formula:
Cl₃Ga MW: 176.08 g/mol Boiling Pt: 201.3 °C Melting Pt: 77.9 °C Density: 2.47 |
MDL Number:
MFCD00011018 CAS Number: 13450-90-3 EINECS: 236-610-0 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...