Gallium phosphide ≥99.999%, PURATREM

Supplier: STREM CHEMICALS INC MS

93-3123-2G 93-3123-10G
100207-032EA 309.97 USD
100207-032 100197-220
Gallium phosphide ≥99.999%, PURATREM
Gallium phosphide
Formula: GaP
MW: 100.7 g/mol
Melting Pt: 1348 °C
Density: 4.1
MDL Number: MFCD00016109
CAS Number: 12063-98-8
EINECS: 235-057-2
Merck Index: 12,04366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR