Iridium(III) acetylacetonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Iridium (III) tris(2,4-pentanedionato-O,O')Iridium (III) 2,4-pentanedionateTris(2,4-pentanedionato-O,O')iridium (III)Iridium(III) 2,4-acetylacetonate

77-1500-5G 77-1500-1G 77-1500-250MG
100201-738EA 135.88 USD
100201-738 100203-298 100203-296
Iridium(III) acetylacetonate ≥98%
Iridium (III) acetylacetonate

ALD, Metal Beta-diketonates, Volatile Precursors for CVD

Formula: C₁₅H₂₁IrO₆
MW: 489.55 g/mol
MDL Number: MFCD00015353
CAS Number: 15635-87-7
EINECS: 239-711-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR