(R,R)-(-)-2,4-Pentanediol ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
(2R,4R)-Pentane-2,4-diol
, (R,R)-(-)-2,4-Dihydroxy pentane
, (2R,4R)-(-)-2,4-Pentanediol
08-2030-1G
08-2030-250MG
101418-450EA
355.26
USD
101418-450
101418-448
(R,R)-(-)-2,4-Pentanediol ≥99%
(R,R)-(-)-2,4-Pentanediol
Formula:
C₅H₁₂O₂ MW: 104.15 g/mol Boiling Pt: 207 °C (1013 hPa) Melting Pt: 48…50 °C Storage Temperature: Ambient |
MDL Number:
MFCD00063893 CAS Number: 42075-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...