1-Ethyl-3-methylimidazolium ethylsulfate ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
EMIM ESU
07-0581-50G
07-0581-250G
101418-416EA
1108.25
USD
101418-416
101418-418
1-Ethyl-3-methylimidazolium ethylsulfate ≥98%
1-Ethyl-3-methylimidazolium ethyl sulphate
Ionic Liquids
Formula:
C₈H₁₆N₂O₄S MW: 236.29 g/mol Melting Pt: < 65 °C Density: 1,2285 |
MDL Number:
MFCD06798189 CAS Number: 342573-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...