Iron(II) bromide hydrate

Supplier: STREM CHEMICALS INC MS
Warning

93-2628-25G 93-2628-100G
100210-292EA 140.12 USD
100210-292 100202-024
Iron(II) bromide hydrate
Iron (II) bromide hydrate
Formula: Br₂Fe·H₂O
MW: 233.67 g/mol
Melting Pt: 27 °C
MDL Number: MFCD00016081
CAS Number: 13463-12-2
EINECS: 232-168-8
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR