Hematite, red ≥99.8% (Fe basis)
Supplier: STREM CHEMICALS INC MS
Synonyms:
Iron (III) oxide, red
93-2617-1000G
93-2617-5000G
93-2617-250G
100203-426EA
526.52
USD
100203-426
100203-424
100204-258
Hematite, red ≥99.8% (Fe basis)
Hematite
Formula:
Fe₂O₃ MW: 159.69 g/mol Melting Pt: 1565 °C Density: 5.26 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011008 CAS Number: 1317-60-8 EINECS: 215-275-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...