Manganese selenide ≥99.9% (Mn basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Manganese(II) selenide

93-2521-5G 93-2521-25G
100204-252EA 155.69 USD
100204-252 100202-016
Manganese selenide ≥99.9% (Mn basis)
Manganese selenide
Formula: MnSe
MW: 133.9 g/mol
Melting Pt: 1460 °C
Density: 5.59 g/cm³ (20 °C)
MDL Number: MFCD00049486
CAS Number: 1313-22-0
EINECS: 215-203-1
UN: 3283
ADR: 6.1,II
Merck Index: 12,05779

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR