Chromium(III) chloride, anhydrous ≥99.9% (Cr basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Chromium trichlorideChromic chloride

93-2442-50G 93-2442-10G
100196-560EA 444.43 USD
100196-560 100210-276
Chromium(III) chloride, anhydrous ≥99.9% (Cr basis)
Chromium (III) chloride
Formula: Cl₃Cr
MW: 158.35 g/mol
Boiling Pt: 1300 °C (subl.)
Melting Pt: ~1150 °C
Density: 2.76
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 12,02278

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR