Titanium(II) hydride ≥95% (99+%-Ti)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Titanium hydrideTitanium dihydride

93-2211-500G 93-2211-25G 93-2211-100G
100203-412EA 563.32 USD
100203-412 100204-230 100199-510
Titanium(II) hydride ≥95% (99+%-Ti)
Titanium (II) hydride
Formula: H₂Ti
MW: 49.9 g/mol
Melting Pt: 450 °C (dec.)
Density: 3.9
MDL Number: MFCD00151301
CAS Number: 7704-98-5
EINECS: 231-726-8
UN: 1871
ADR: 4.1,II
Merck Index: 14,09473

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR