Calcium-2-ethylhexanoate 40% in 2-ethylhexanoic acid (3-8% Ca), Superconductor Grade
Supplier: STREM CHEMICALS INC MS
Synonyms:
2-Ethylhexanoic acid calcium salt
, Calcium 2-ethylhexanoate
, 2-Ethylcaproic acid calcium salt
93-2014-100G
93-2014-25G
100208-128EA
890.28
USD
100208-128
100208-130
Calcium-2-ethylhexanoate 40% in 2-ethylhexanoic acid (3-8% Ca), Superconductor Grade
Calcium 2-ethylhexanoate
Formula:
C₁₆H₃₀CaO₄ MW: 326.49 g/mol |
MDL Number:
MFCD00014001 CAS Number: 136-51-6 EINECS: 205-249-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...