Calcium chromate ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

93-2012-100G 93-2012-500G
100201-958EA 184 USD
100201-958 100198-996
Calcium chromate ≥95%
Calcium chromate
Formula: CaCrO₄
MW: 156.07 g/mol
Melting Pt: 2170 °C
Density: 3.12 g/cm³ (20 °C)
MDL Number: MFCD00049403
CAS Number: 13765-19-0
EINECS: 237-366-8
UN: 3087
ADR: 5.1,II
Merck Index: 12,01700

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR