Calcium acetate monohydrate ≥98%
Supplier: STREM CHEMICALS INC MS
Synonyms:
Calcium di(acetate) monohydrate
, Calcium acetate hydrate
, Acetic acid calcium salt monohydrate
93-2001-250G
93-2001-1000G
100196-532EA
256.18
USD
100196-532
100201-956
Calcium acetate monohydrate ≥98%
Calcium acetate monohydrate
Formula:
(H₃CCOO)₂Ca·1H₂O MW: 176.18 g/mol Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 EINECS: 200-540-9 Merck Index: 13,01645 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...