Triphenyl phosphate ≥98%

Supplier: STREM CHEMICALS INC MS

Warning

Synonyms: TPP

93-1562.500
100203-398EA 120.31 USD InStock
100203-398
Triphenyl phosphate ≥98%
Triphenyl phosphate
Formula: C₁₈H₁₅O₄P
MW: 326.29 g/mol
Boiling Pt: 244°/10 mm
Melting Pt: 49-51°
Density: 1.33 g/cm³ (20 °C)
Flash Pt: 428°F
Storage Temperature: Ambient
MDL Number: MFCD00003031
CAS Number: 115-86-6
EINECS: 204-112-2
UN: 3077
ADR: 9,III
Merck Index: 12,09872

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR