Vinyltriethoxysilane ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Triethoxy(vinyl)silane
93-1471-500G
93-1471-100G
100207-782EA
62.28
USD
100207-782
100201-066
Vinyltriethoxysilane ≥97%
Vinyltriethoxysilane
Formula:
C₈H₁₈O₃Si MW: 190.31 g/mol Boiling Pt: 161 °C Density: 0,9050 (20°) Flash Pt: 94 °F Storage Temperature: Ambient |
MDL Number:
MFCD00009063 CAS Number: 78-08-0 EINECS: 201-081-7 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...