Tetraiodosilane ≥99.9% (Si basis)
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1447-2G
93-1447-10G
100203-384EA
121.72
USD
100203-384
100208-110
Tetraiodosilane ≥99.9% (Si basis)
Tetraiodosilane
Formula:
I₄Si MW: 535.7 g/mol Boiling Pt: 287.5 °C Melting Pt: 120.5 °C Density: 4.198 |
MDL Number:
MFCD00054041 CAS Number: 13465-84-4 EINECS: 236-706-2 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...