Methyltriethoxysilane ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Triethoxy(methyl)silane
93-1418-100G
93-1418-500G
100201-914EA
67.94
USD
100201-914
100198-308
Methyltriethoxysilane ≥98%
Methyltriethoxysilane
Formula:
C₇H₁₈O₃Si MW: 178.3 g/mol Boiling Pt: 143 °C Melting Pt: –50 °C Density: 0,896 Flash Pt: 75 °F Storage Temperature: Refrigerator |
MDL Number:
MFCD00009066 CAS Number: 2031-67-6 EINECS: 217-983-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...