Ethyltrichlorosilane ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Trichloro(ethyl)silaneEthyltrichlorosilane

93-1411-100G 93-1411-500G
100205-330EA 134.46 USD
100205-330 100196-502
Ethyltrichlorosilane ≥98%
Ethyltrichlorosilane
Formula: C₂H₅Cl₃Si
MW: 163.51 g/mol
Boiling Pt: 98.8 °C
Melting Pt: –105.6 °C
Density: 1.238
Flash Pt: 14 °F
MDL Number: MFCD00000485
CAS Number: 115-21-9
EINECS: 204-072-6
UN: 1196
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR