Aluminium carbide ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Aluminum carbideTetraaluminium tricarbide

93-1386-100G 93-1386-25G
100205-320EA 920 USD
100205-320 100196-498
Aluminium carbide ≥98%
Aluminium carbide
Formula: C₃Al₄
MW: 143.96 g/mol
Melting Pt: 2100 °C
Density: 2.36
MDL Number: MFCD00003425
CAS Number: 1299-86-1
EINECS: 215-076-2
UN: 1394
ADR: 4.3,II
Merck Index: 12,00345

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR