Trimethylaluminium ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Trimethylaluminum
93-1360-100G
93-1360-225G
100198-978EA
390.65
USD
100198-978
100201-060
Trimethylaluminium ≥98%
Trimethylaluminium
Metal Alkyls, Volatile Precursors for CVD
Formula:
C₃H₉Al MW: 72.09 g/mol Boiling Pt: 20 °C (8 mmHg) Melting Pt: 15.4 °C Density: 0.752 (20°) Flash Pt: 1.4 °F |
MDL Number:
MFCD00008252 CAS Number: 75-24-1 EINECS: 200-853-0 UN: 3394 ADR: 4.2,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...