Aluminium sulfide ≥99% (Al basis) (1/4'' and down pieces)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Dialluminium trisulphideDialluminium trisulfideAluminum sulphide

93-1353-25G
100201-896EA 527.94 USD
100201-896
Aluminium sulfide ≥99% (Al basis) (1/4'' and down pieces)
Aluminium sulphide
Formula: Al₂S₃
MW: 150.16 g/mol
Boiling Pt: 1500 °C (1013 hPa)
Melting Pt: 1100 °C
Density: 2.02
MDL Number: MFCD00014162
CAS Number: 1302-81-4
EINECS: 215-109-0
UN: 3132
ADR: 4.3,I
Merck Index: 12,00382

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR