Aluminium ≥99.7%, Powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Aluminum
93-1344-500G
93-1344-2000G
100210-182EA
380.74
USD
100210-182
100207-776
Aluminium ≥99.7%, Powder
Aluminium
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2056 °C Melting Pt: 660 °C Density: 2,702 |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...