Aluminium ≥99%, Powder
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Aluminum
93-1316-500G
93-1316-2000G
100205-310EA
92
USD
100205-310
100200-512
Aluminium ≥99%, Powder
Aluminium
Formula:
Al MW: 26.98 g/mol Boiling Pt: 2056 °C Melting Pt: 660 °C Density: 2,702 |
MDL Number:
MFCD00134029 CAS Number: 7429-90-5 EINECS: 231-072-3 UN: 1396 ADR: 4.3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...