Aluminium acetylacetonate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Aluminium tris(2,4-pentanedionato-O,O')
, Aluminum acetylacetonate
, Aluminum 2,4-pentanedionate
, Tris(2,4-pentanedionato-O,O')aluminium
, Aluminium 2,4-pentanedionate
93-1302-100G
93-1302-500G
100207-774EA
169.85
USD
100207-774
100210-176
Aluminium acetylacetonate ≥99%
Aluminium acetylacetonate
Metal Beta-diketonates, Volatile Precursors for CVD
Formula:
C₁₅H₂₁AlO₆ MW: 324.31 g/mol Boiling Pt: 320 °C/ 150 °C (1 mmHg) (dec./ subl.) Melting Pt: 192…193 °C Density: 1.204 g/cm³ (20 °C) |
MDL Number:
MFCD00000013 CAS Number: 13963-57-0 EINECS: 237-741-6 UN: 3467 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...