Magnesium perchlorate hexahydrate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

93-1248-100G 93-1248-500G
100197-680EA 35.38 USD
100197-680 100206-928
Magnesium perchlorate hexahydrate ≥99%
Magnesium perchlorate hexahydrate
Formula: Cl₂MgO₈·6H₂O
MW: 331.3 g/mol
Melting Pt: 185…190 °C
Density: 1.98
MDL Number: MFCD00149778
CAS Number: 13446-19-0
EINECS: 233-108-3
UN: 1475
ADR: 5.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR