Magnesium oxide ≥98% -325 mesh
|
Warning
|
Synonyms:
Magnesia usta
93-1244-1000G
100204-160EA
109.33
USD
100204-160
Magnesium oxide ≥98% -325 mesh
Magnesium oxide
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 °C Melting Pt: 2800 °C Density: 3,58 |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...