Sodium perchlorate, anhydrous ≥98% ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-1169-500G
93-1169-100G
100211-204EA
198.15
USD
100211-204
100204-152
Sodium perchlorate, anhydrous ≥98% ACS
Sodium perchlorate
Formula:
ClNaO₄ MW: 122.44 g/mol Melting Pt: 482 °C (dec.) Density: 2.02 Storage Temperature: Ambient |
MDL Number:
MFCD00003480 CAS Number: 7601-89-0 EINECS: 231-511-9 UN: 1502 ADR: 5.1,II Merck Index: 12,08798 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...