Sodium molybdate(VI), anhydrous ≥98%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
di-Sodium molybdate
, Molybdic acid sodium salt
, Sodium molybdenum oxide
, Sodium molybdate
, Molybdic acid, sodium salt
93-1160-500G
93-1160-100G
100200-044EA
267.51
USD
100200-044
100200-042
Sodium molybdate(VI), anhydrous ≥98%
Sodium molybdate(VI)
Formula:
MoNa₂O₄ MW: 205.92 g/mol Melting Pt: 687 °C Density: 3.28 |
MDL Number:
MFCD00003486 CAS Number: 7631-95-0 EINECS: 231-551-7 Merck Index: 13,08718 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...