Sodium methoxide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium methylate
, Sodium methanolate
93-1159-1000G
93-1159-500G
100206-908EA
168.43
USD
100206-908
100196-458
Sodium methoxide ≥98%
Sodium methoxide
Alkoxide
Formula:
CH₃NaO MW: 54.02 g/mol Melting Pt: 127 °C Density: 1.3 g/cm³ (20 °C) |
MDL Number:
MFCD00012179 CAS Number: 124-41-4 EINECS: 204-699-5 UN: 1431 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...