Sodium ethoxide ≥96%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Sodium ethylate
, Sodium ethanolate
93-1139-50G
93-1139-250G
100204-142EA
32.55
USD
100204-142
100204-140
Sodium ethoxide ≥96%
Sodium ethoxide
Alkoxide
Formula:
C₂H₅NaO MW: 68.05 g/mol Melting Pt: 260 °C Density: 0.868 g/cm³ (20 °C) |
MDL Number:
MFCD00012417 CAS Number: 141-52-6 EINECS: 205-487-5 UN: 3206 ADR: 4.2,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...