Sodium cyanide ≥98%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
93-1132-500G
93-1132-2000G
100200-036EA
114
USD
100200-036
100200-508
Sodium cyanide ≥98%
Sodium cyanide
Formula:
CNNa MW: 49.01 g/mol Boiling Pt: 1496 °C Melting Pt: 563.7 °C Density: 1.595 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003523 CAS Number: 143-33-9 EINECS: 205-599-4 UN: 1689 ADR: 6.1,I Merck Index: 12,08750 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...