EDTA disodium salt dihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Ethylenediaminetetra-acetic acid disodium salt dihydrate
, Edetate disodium dihydrate
, Ethylenediaminetetraacetic acid, disodium salt, dihydrate
, Kestranal® 2S 2 H2O
, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate
, Titriplex III
, Kestranal® 2S dihydrate
, Edetic acid disodium salt dihydrate
, Disodium dihydrogen ethylenediaminetetraacetate dihydrate
93-1103-1000G
93-1103-250G
100210-850EA
210.89
USD
100210-850
100201-854
EDTA disodium salt dihydrate ≥99%
EDTA disodium salt dihydrate
Formula:
C₁₀H₁₄N₂Na₂O₈·2H₂O MW: 372.24 g/mol Melting Pt: 255 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150037 CAS Number: 6381-92-6 EINECS: 205-358-3 Merck Index: 13,03543 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...