Nitronium hexafluoroantimonate ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nitryl hexafluoroantimonate

93-0738-10G
100200-310EA 188.25 USD
100200-310
Nitronium hexafluoroantimonate ≥97%
Nitronium hexafluoroantimonate
Formula: F₆NO₂Sb
MW: 281.76 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042551
CAS Number: 17856-92-7
UN: 2923
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR