Lithium tetrahydroborate ≥95%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
Synonyms:
Lithium borohydride
, Lithium tetrahydridoborate
93-0397-1G
93-0397-5G
93-0397-25G
100209-404EA
353.85
USD
100209-404
100196-428
100198-934
Lithium tetrahydroborate ≥95%
Lithium tetrahydroborate
Formula:
H₄BLi Boiling Pt: 66 °C (1013 hPa) Melting Pt: 279 °C (dec.) Density: 0.66 Storage Temperature: Ambient |
MDL Number:
MFCD00011088 CAS Number: 16949-15-8 EINECS: 241-021-7 UN: 1413 ADR: 4.3,I Merck Index: 13,05547 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...