Lithium tantalate ≥99.998% (Ta basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lithium tantalum trioxide
, Lithium tantalum oxide
93-0384-2G
93-0384-10G
100205-252EA
67.94
USD
100205-252
100198-282
Lithium tantalate ≥99.998% (Ta basis), PURATREM
Lithium tantalate
Formula:
LiO₃Ta MW: 235.89 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016174 CAS Number: 12031-66-2 EINECS: 234-757-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...