Lithium dihydrogen orthophosphate ≥97%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
Synonyms:
Lithium phosphate monobasic
, Lithium dihydrogenorthophosphate
, Lithium dihydrogenphosphate
, Lithium dihydrogen phosphate
93-0383-500G
93-0383-100G
100205-250EA
97.66
USD
100205-250
100199-472
Lithium dihydrogen orthophosphate ≥97%
Lithium dihydrogen orthophosphate
Formula:
H₂LiO₄P MW: 103.93 g/mol Density: 2,461 |
MDL Number:
MFCD00016188 CAS Number: 13453-80-0 EINECS: 236-633-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...