Lithium iodide trihydrate ≥99%
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-0381-250G
93-0381-50G
100203-348EA
144.37
USD
100203-348
100200-024
Lithium iodide trihydrate ≥99%
Lithium iodide trihydrate
Formula:
ILi·3H₂O MW: 187.89 g/mol Boiling Pt: 1171 °C (1013 hPa) Melting Pt: 70 °C Density: 3,43 Storage Temperature: Ambient |
MDL Number:
MFCD00149770 CAS Number: 7790-22-9 EINECS: 233-822-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...